accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 100-47-0

IUPAC Name: benzonitrile
Synonyms: benzonitrile benzonitrile (substance) fenylkyanid benzoic acid nitrile C6H5-CN hsdb 45
Molecular Weight (g/mol): 103.12
Molecular Formula: C7H5N
InChi Key: JFDZBHWFFUWGJE-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (6)
Boiling Point 
  • (7)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (5)
  • (3)
Grade 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (10)
  • (7)
  • (6)
  • (7)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
  • (1)