Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzonitrile, for HPLC

Catalog No. AC446911000
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446911000 100 mL Glass bottle
AC446910010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446911000 Supplier Thermo Scientific Chemicals Supplier No. 446911000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 100-47-0 | C7H5N | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-47-0
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.12
InChI Key JFDZBHWFFUWGJE-UHFFFAOYSA-N
Synonym phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
PubChem CID 7505
ChEBI CHEBI:27991
IUPAC Name benzonitrile
SMILES C1=CC=C(C=C1)C#N

Specifications

Melting Point -13.0°C
Density 1.0100g/mL
Boiling Point 188.0°C to 191.0°C
Flash Point 71°C
Packaging Glass bottle
Refractive Index 1.5270 to 1.5290
Linear Formula C6H5CN
Quantity 100 mL
Merck Index 15,1099
Specific Gravity 1.01
Solubility Information Solubility in water: 10g/L (100°C). Other solubilities: misicble with most common organic solvents
Viscosity 1.24 mPa.s (25°C)
Formula Weight 103.12
Grade HPLC
Chemical Name or Material Benzonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 202-855-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.