accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-50-5

IUPAC Name: cyclohex-3-ene-1-carbaldehyde
Synonyms: 3-cyclohexene-1-carboxaldehyde
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: DCFDVJPDXYGCOK-UHFFFAOYNA-N
SMILES: O=CC1CCC=CC1
Boiling Point 
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)