Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclohexene-1-carboxaldehyde, 98%, Thermo Scientific™

Catalog No. AAL04894AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL04894AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL04894AD Supplier Thermo Scientific Chemicals Supplier No. L04894AD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-50-5
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD00001572
InChI Key DCFDVJPDXYGCOK-UHFFFAOYNA-N
Synonym 3-cyclohexene-1-carboxaldehyde, 1,2,3,6-tetrahydrobenzaldehyde, 4-formylcyclohexene, cyclohexene-4-carboxaldehyde, 1-formyl-3-cyclohexene, 3-cyclohexen-1-aldehyde, 1-cyclohexene-4-carboxaldehyde, 1,2,5,6-tetrahydrobenzaldehyde, cyclohex-3-enecarbaldehyde, tetrahydrobenzaldehyde
PubChem CID 7508
IUPAC Name cyclohex-3-ene-1-carbaldehyde
SMILES O=CC1CCC=CC1

Specifications

Melting Point 1°C to 3°C
Boiling Point 163°C to 164°C
Flash Point 57°C (134°F)
Quantity 50 mL
UN Number UN2498
Beilstein 774001
Formula Weight 110.16
Percent Purity 98%
Chemical Name or Material 3-Cyclohexene-1-carboxaldehyde
Hazard Category Danger
Hazard Statement GHS H Statement
H311-H226-H332-H319-H335
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 1,2,3,6-TETRAHYDROBENZALDEHYDE
RTECSNumber GW2800000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.