accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CN

CAS RN 100-62-9

IUPAC Name: N-phenylmethanimine
Synonyms: N-phenylmethanimine
Molecular Weight (g/mol): 105.14
Molecular Formula: C7H7N
InChi Key: VULHYRAYXYTONQ-UHFFFAOYSA-N
SMILES: C=NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results