Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyleneaniline,Trimer, MP Biomedicals

Catalog No. ICN216968 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216968 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216968 Supplier MP Biomedicals, Inc Supplier No. 0521696880
Add to Cart
Add to Cart

Chemical Identifiers

CAS 100-62-9
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
InChI Key VULHYRAYXYTONQ-UHFFFAOYSA-N
Synonym n-methyleneaniline, benzenamine, n-methylene, methylenanilin, n-methylene aniline, n-methylideneaniline, methanimine, n-phenyl, benzenamine,n-methylene, methyleneaniline,trimer
PubChem CID 66014
IUPAC Name N-phenylmethanimine
SMILES C=NC1=CC=CC=C1

Specifications

Quantity 100 g
Formula Weight 315.42
Chemical Name or Material Methyleneaniline, Trimer
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.