accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 100-66-3

IUPAC Name: anisole
Synonyms: anisole fema number 2097 anizol hsdb 44
Molecular Weight (g/mol): 108.14
Molecular Formula: C7H8O
InChi Key: RDOXTESZEPMUJZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (10)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (4)
  • (6)
Grade 
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (10)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (8)
  • (1)