Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anisole, ReagentPlus™, 99%, Solstice

Catalog No. 60044724 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-724 2.5 L Glass Bottle
60-044-723 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-724 Supplier Solstice Supplier No. 1232262.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-66-3
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00008354
InChI Key RDOXTESZEPMUJZ-UHFFFAOYSA-N
Synonym methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
PubChem CID 7519
ChEBI CHEBI:16579
IUPAC Name anisole
SMILES COC1=CC=CC=C1

Specifications

Melting Point -37°C
Color Colorless
Density 0.995g/mL at 25°C
Boiling Point 154°C
Flash Point 51°C (123.8°F)
Packaging Glass Bottle
Refractive Index n20/D 1.516(lit.)
Linear Formula CH3OC6H5
Quantity 2.5 L
UN Number UN2222
Beilstein 506892
Vapor Density 3.7 (vs air)
Formula Weight 108.14g/mol
Percent Purity 99%
Grade ReagentPlus
Physical Form Liquid
Chemical Name or Material Anisole
Show More Show Less
Hazard Statement P210-P370 + P378
Precautionary Statement H226
DOTInformation 3/PGIII
EINECSNumber 202-876-1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.