accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100047-36-7

IUPAC Name: 4-aminopyridine-2-carboxylic acid
Synonyms: 4-aminopyridine-2-carboxylic acid
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: JRZBTJVSAANBEV-UHFFFAOYSA-N
SMILES: NC1=CC(=NC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)