Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminopyridine-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AC455772500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455772500 250 mg
AC455770010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455772500 Supplier Thermo Scientific Chemicals Supplier No. 455772500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100047-36-7
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.12
InChI Key JRZBTJVSAANBEV-UHFFFAOYSA-N
Synonym 4-aminopicolinic acid, 4-aminopyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 4-amino, 4-amino-2-pyridinecarboxylic acid, 4-amino-pyridine-2-carboxylic acid, 4-aminopicolinicacid, 4-amino-picolinsaure, pubchem22241, 2-carboxy-4-aminopyridine, picolinic acid, 4-amino
PubChem CID 2050132
IUPAC Name 4-aminopyridine-2-carboxylic acid
SMILES C1=CN=C(C=C1N)C(=O)O

Specifications

Melting Point 319.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 250 mg
Formula Weight 138.12
Percent Purity 97%
Chemical Name or Material 4-Aminopyridine-2-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.