accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100063-22-7

IUPAC Name: methyl 3-amino-5-phenylthiophene-2-carboxylate
Synonyms: methyl 3-amino-5-phenylthiophene-2-carboxylate
Molecular Weight (g/mol): 233.29
Molecular Formula: C12H11NO2S
InChi Key: QESSCNMSOLRYBO-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=C(S1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results