Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle3-amino-5-phenylthiophene-2-carboxylate, 97%, Thermo Scientific™

Catalog No. GK00498EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
GK00498EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GK00498EA Supplier Thermo Scientific Chemicals Supplier No. GK00498EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100063-22-7
Molecular Formula C12H11NO2S
Molecular Weight (g/mol) 233.285
MDL Number MFCD00068161
InChI Key QESSCNMSOLRYBO-UHFFFAOYSA-N
Synonym methyl 3-amino-5-phenyl-2-thiophenecarboxylate, 2-thiophenecarboxylic acid, 3-amino-5-phenyl-, methyl ester, methyl-3-amino-5-phenylthiophene-2-carboxylate, methyl 3-amino-5-4-phenyl thiophene-2-carboxylate, 3-amino-5-phenyl-thiophene-2-carboxylic acid methyl ester, maybridge1_003977, acmc-2097mh, methyl-3-amino-5-phenylthiophene-carboxylate, 3-amino-2-methoxycarbonyl-5-phenylthiophene, methyl 3-amino-5-phenyl-thiophene-2-carboxylate
PubChem CID 700562
IUPAC Name methyl 3-amino-5-phenylthiophene-2-carboxylate
SMILES COC(=O)C1=C(C=C(S1)C2=CC=CC=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 233.29
Percent Purity 97%
Chemical Name or Material Methyle3-amino-5-phenylthiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.