accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1001-89-4

IUPAC Name: 2-chloroheptane
Synonyms: 2-chloroheptane
Molecular Weight (g/mol): 134.65
Molecular Formula: C7H15Cl
InChi Key: PTSLUOSUHFGQHV-UHFFFAOYNA-N
SMILES: CCCCCC(C)Cl
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results