accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10010-67-0

IUPAC Name: sodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate
Synonyms: sodium pipes|pipes monosodium|pipes monosodium salt|monosodium pipes
Molecular Weight (g/mol): 324.34
Molecular Formula: C8H17N2NaO6S2
InChi Key: OGGAIRCLBMGXCZ-UHFFFAOYSA-M
SMILES: [Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)