Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIPES, Monosodium Salt, Biological Buffer, High Purity, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18612390 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-390 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-612-390 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P10511KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10010-67-0 | C8H17N2NaO6S2 | 324.34 g/mol

Chemical Identifiers

CAS 10010-67-0
Molecular Formula C8H17N2NaO6S2
Molecular Weight (g/mol) 324.34
MDL Number MFCD00065472
InChI Key OGGAIRCLBMGXCZ-UHFFFAOYSA-M
IUPAC Name sodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

CAS Max % 100%
Physical Form Solid
Quantity 1 kg
Solution Type Buffer
Loss on Drying 5%
Packaging Poly Bottle
Percent Purity 98.5%
Grade High Purity
Chemical Name or Material 1,4-Piperazinediethanesulfonic Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.