CAS RN 1001478-90-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Choline Kinase-a Inhibitor, CK37
The Choline Kinase-α Inhibitor, CK37, also referenced under CAS 1001478-90-5, controls the biological activity of Choline Kinase-α. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.