accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100173-92-0

IUPAC Name: dibutyl(2-{[2-(dibutylamino)ethyl]amino}ethyl)amine
Synonyms: dibutyl(2-{[2-(dibutylamino)ethyl]amino}ethyl)amine
Molecular Weight (g/mol): 327.6
Molecular Formula: C20H45N3
InChi Key: LLHVBJQIWZHCCV-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)CCNCCN(CCCC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results