Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N″,N″-Tetrabutyldiethylenetriamine 98.0+%, TCI America™
SDP

Catalog No. T18435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18435G Supplier TCI America Supplier No. T18435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100173-92-0
Molecular Formula C20H45N3
Molecular Weight (g/mol) 327.60
MDL Number MFCD02093504
InChI Key LLHVBJQIWZHCCV-UHFFFAOYSA-N
Synonym Bis[2-(dibutylamino)ethyl]amine
PubChem CID 17793884
IUPAC Name dibutyl(2-{[2-(dibutylamino)ethyl]amino}ethyl)amine
SMILES CCCCN(CCCC)CCNCCN(CCCC)CCCC

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 327.60
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N,N,N′′,N′′-Tetrabutyldiethylenetriamine
EINECSNumber (7)-0005
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.