accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1002-67-1

IUPAC Name: 1-ethoxy-2-(2-methoxyethoxy)ethane
Synonyms: 1-ethoxy-2-(2-methoxyethoxy)ethane
Molecular Weight (g/mol): 148.2
Molecular Formula: C7H16O3
InChi Key: CNJRPYFBORAQAU-UHFFFAOYSA-N
SMILES: CCOCCOCCOC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results