accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCl

CAS RN 1002-69-3

IUPAC Name: 1-chlorodecane
Synonyms: 1-chlorodecane
Molecular Weight (g/mol): 176.73
Molecular Formula: C10H21Cl
InChi Key: ZTEHOZMYMCEYRM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCl
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)