Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorodecane 97.0+%, TCI America™
SDP

Catalog No. C060025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C060025ML 25 mL
C0600500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C060025ML Supplier TCI America Supplier No. C060025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1002-69-3
Molecular Formula C10H21Cl
Molecular Weight (g/mol) 176.73
MDL Number MFCD00000956
InChI Key ZTEHOZMYMCEYRM-UHFFFAOYSA-N
Synonym decyl chloride, decane, 1-chloro, n-decyl chloride, decane, chloro, unii-l0hsc44e5x, decyl chloride mixed isomers, chlorodecane, l0hsc44e5x, chlorodecane mixed isomers, 1-chlorodecane
PubChem CID 13848
IUPAC Name 1-chlorodecane
SMILES CCCCCCCCCCCl

Specifications

Melting Point -34°C
Color Yellow
Boiling Point 223°C
Quantity 25 mL
UN Number 3082
Formula Weight 176.73
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorodecane
EINECSNumber (2)-0066&(2)-0068
RTECSNumber HD7050000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.