accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10022-28-3

IUPAC Name: 1,1-dimethoxyoctane
Synonyms: octanal dimethyl acetal
Molecular Weight (g/mol): 174.28
Molecular Formula: C10H22O2
InChi Key: BZOOCKAFKVYAOZ-UHFFFAOYSA-N
SMILES: CCCCCCCC(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results