CAS RN 10025-69-1
Tin(II) chloride dihydrate, 98%
CAS: 10025-69-1 Molecular Formula: Cl2Sn·2H2O MDL Number: MFCD00149863 Synonym: Tin (2+) dihydrate dichloride IUPAC Name: Tin (2+) dihydrate dichloride
Tin(II) chloride dihydrate, 98+%, ACS reagent
CAS: 10025-69-1 Molecular Formula: Cl2Sn·2H2O Molecular Weight (g/mol): 225.63 MDL Number: MFCD00149863 Synonym: Stannous chloride dihydrate
Tin(II) chloride dihydrate, 98+%
CAS: 10025-69-1 Molecular Formula: Cl2Sn·2H2O Molecular Weight (g/mol): 225.63 MDL Number: MFCD00149863 Synonym: Stannous chloride dihydrate
Stannous Chloride Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 10025-69-1 Molecular Formula: Cl2Sn · 2 H2O MDL Number: MFCD00149863 Synonym: Tin (II) Chloride Dihydrate
Tin(II) chloride dihydrate, ACS, 98.0-103.0%
CAS: 10025-69-1 Molecular Formula: SnCl2·2H2O MDL Number: MFCD00149863
Stannous Chloride, Dihydrate, ACS Grade, 98.0% to 103.0%, LabChem™
CAS: 10025-69-1 Synonym: Tin Chloride
Tin(II) chloride dihydrate, 97%
CAS: 10025-69-1 Molecular Formula: Cl2Sn·2H2O Molecular Weight (g/mol): 225.63 MDL Number: MFCD00149863 Synonym: Stannous chloride dihydrate
Tin(II) chloride dihydrate, low in Mercury, suitable for AAS
CAS: 10025-69-1 Molecular Formula: Cl2Sn·2H2O Molecular Weight (g/mol): 225.63 Synonym: Stannous chloride dihydrate
Stannous Chloride, Dihydrate, Technical, 98.0-103.0%, Spectrum™ Chemical
CAS: 10025-69-1
Stannous Chloride, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 10025-69-1 Molecular Formula: SnCl2·2H2O
Tin(II) chloride dihydrate, For AAS, ≥96.0% (RT), MilliporeSigma™ Supelco™
CAS: 10025-69-1 Molecular Formula: Cl2H4O2Sn Molecular Weight (g/mol): 225.64 MDL Number: MFCD00149863 InChI Key: FWPIDFUJEMBDLS-UHFFFAOYSA-L Synonym: Stannous chloride dihydrate IUPAC Name: λ²-tin(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Sn++]
Stannous Chloride, Dihydrate, 98.0 to 101.0%, ACS, MilliporeSigma™
CAS: 10025-69-1 Molecular Formula: SnCl2·2H2O Synonym: Tin(II) Chloride, Dihydrate
Stannous Chloride, Dihydrate, Crystal, Reagent, ACS, 98-103%, Spectrum™ Chemical
CAS: 10025-69-1 Molecular Formula: Cl2H4O2Sn Molecular Weight (g/mol): 225.64 InChI Key: FWPIDFUJEMBDLS-UHFFFAOYSA-L IUPAC Name: λ2-tin(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Sn++]
Stannous Chloride Dihydrate, MP Biomedicals
CAS: 10025-69-1 Molecular Formula: SnCl2 · 2H2O
Tin(II) Chloride Dihydrate, For Analysis, EMSURE™Test Kit, For water determination, According to Karl Fischer, Apura™, MilliporeSigma™
CAS: 10025-69-1 Molecular Formula: SnCl2·2H2O Synonym: Tin Dichloride
- 1
- 2