accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OClClClEr

CAS RN 10025-75-9

IUPAC Name: erbium(3+) hexahydrate trichloride
Synonyms: erbium(3+) hexahydrate trichloride
Molecular Weight (g/mol): 381.7
Molecular Formula: Cl3ErH12O6
InChi Key: UJBPGOAZQSYXNT-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Er+3]
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)