accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClPClClO

CAS RN 10025-87-3

IUPAC Name: phosphoroyl trichloride
Synonyms: phosphorus oxychloride phosphoric trichloride phosphoroxidchlorid phosphorylchlorid pocl3 phosphoroxychlorid phosphoryl chloride phosphorus(V) trichloride oxide phosphortrichloridoxid trichlorure de phosphoryle
Molecular Weight (g/mol): 153.32
Molecular Formula: Cl3OP
InChi Key: XHXFXVLFKHQFAL-UHFFFAOYSA-N
SMILES: ClP(Cl)(Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (4)
  • (3)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (5)
  • (4)
  • (3)
  • (2)
  • (5)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (4)
  • (3)
  • (7)
  • (5)
  • (4)