Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus Oxychloride, 98%, Spectrum™ Chemical
SDP

Catalog No. 18612354 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
20 kg
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-354 250 g Steel Can
18-612-351 5 g Steel Can
18-612-352 1 kg Steel Can
18-612-353 20 kg Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-354 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1115250GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10025-87-3

Specifications

CAS Max % 1
Physical Form Liquid
Quantity 250 g
Molecular Weight (g/mol) 153.33
Percent Purity 98%
Grade Reagent
Packaging Steel Can
Chemical Name or Material Phosphorus Oxychloride

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.