accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10026-04-7

IUPAC Name: tetrachlorosilane
Synonyms: silicon tetrachloride
Molecular Weight (g/mol): 169.89
Molecular Formula: Cl4Si
InChi Key: FDNAPBUWERUEDA-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (11)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
Physical Form 
  • (11)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (8)
  • (11)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (11)
  • (5)