Learn More
Silicon(IV) chloride, 99.8+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 58.0°C |
| Melting Point | -70.0°C |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 50 mL |
| Packaging | Glass bottle |
| Refractive Index | 1.413 |
| Specific Gravity | 1.48 |
| Assay Percent Range | 99.8+% |
| Density | 1.4800g/mL |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Toxic if swallowed. Toxic if inhaled. Causes severe skin burns and eye damage. Reacts violently with water. |
| Precautionary Statement | GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. |
| EINECSNumber | 233-054-0 |
| RTECSNumber | VW0525000 |
| TSCA | TSCA |
| Recommended Storage | Product may develop some turbidity or precipitate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.