accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClZr

CAS RN 10026-11-6

IUPAC Name: zirconium(4+) tetrachloride
Synonyms: zirconium(IV) chloride
Molecular Weight (g/mol): 233.02
Molecular Formula: Cl4Zr
InChi Key: DUNKXUFBGCUVQW-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Zr+4]
Quantity 
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (7)
Percent Purity 
  • (3)
  • (2)
  • (4)
  • (2)
Physical Form 
  • (4)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (7)
  • (8)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (7)
  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)