Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zirconium(IV) chloride

Catalog No. 35740510G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-740-510G 10 g
35-740-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-740-510G Supplier Sigma Aldrich Supplier No. 35740510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zirconium (IV) chloride acts as an efficient Lewis acid catalyst for various reactions; such as:electrophilic addition reaction of indole with aldehydes/ketones to form bis(indolyl)methanes. electrophilic amination of activated arenes,transthioacetylization of acetals,deoxygenation of heterocyclic-N-oxides,reduction of nitro compounds,conversion of carbonyl compounds to 1,3-oxathiolanes,Biginelli reaction for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones,direct amide coupling of non activated carboxylic acids and amines,synthesis of quinoxalines and pyrido[2,3-b]pyrazines,deprotection of t-butyldimethylsilyl (TBDMS) ethers.Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the correspondingα,α-dimethylamines.

Specifications

CAS 10026-11-6
Density 2.8 g/mL (at 25°C)
Molecular Formula Cl4Zr
Linear Formula ZrCl4
MDL Number MFCD00011306
Quantity 10 g
Synonym Tetrachlorozirconium; Zirconium tetrachloride
Molecular Weight (g/mol) 233.04
Percent Purity ≥99.9% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.