CAS RN 10028-24-7
Sodium Phosphate Dibasic Dihydrate (USP/FCC/EP/BP), Fisher Chemical™
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate dihydrate,disodium phosphate dihydrate,sodium phosphate dibasic dihydrate,di-sodium hydrogen phosphate dihydrate,phosphoric acid, disodium salt, dihydrate,sodium hydrogenphosphate dihydrate,sodium phosphate, dibasic, dihydrate,disodium hydrogenphosphate dihydrate,dinatrii phosphas dihydricus,2na.hpo4.2h2o PubChem CID: 6451167 IUPAC Name: disodium dihydrate hydrogen phosphate SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
Sodium phosphate dibasic dihydrate, 99+%, for HPLC
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate dihydrate,disodium phosphate dihydrate,sodium phosphate dibasic dihydrate,di-sodium hydrogen phosphate dihydrate,phosphoric acid, disodium salt, dihydrate,sodium hydrogenphosphate dihydrate,sodium phosphate, dibasic, dihydrate,disodium hydrogenphosphate dihydrate,dinatrii phosphas dihydricus,2na.hpo4.2h2o PubChem CID: 6451167 IUPAC Name: disodium;hydrogen phosphate;dihydrate SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
Sodium phosphate dibasic dihydrate, 98+%, for analysis
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate dihydrate,disodium phosphate dihydrate,sodium phosphate dibasic dihydrate,di-sodium hydrogen phosphate dihydrate,phosphoric acid, disodium salt, dihydrate,sodium hydrogenphosphate dihydrate,sodium phosphate, dibasic, dihydrate,disodium hydrogenphosphate dihydrate,dinatrii phosphas dihydricus,2na.hpo4.2h2o PubChem CID: 6451167 IUPAC Name: disodium;hydrogen phosphate;dihydrate SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
Sodium Phosphate, Dibasic, Dihydrate, FCC, 98%, Spectrum™ Chemical
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L IUPAC Name: disodium dihydrate hydrogen phosphate SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
Sodium phosphate, dibasic dihydrate, 98%, extra pure
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L IUPAC Name: disodium dihydrate hydrogen phosphate SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
Sodium Phosphate, Dibasic, Dihydrate, USP, EP, bioCERTIFIED™, 10 kg, Spectrum Chemical
CAS: 10028-24-7
Sodium Phosphate, Dibasic, Dihydrate, USP, EP, bioCERTIFIED™, 1 kg, Spectrum Chemical
CAS: 10028-24-7 Molecular Weight (g/mol): 177.99
Sodium phosphate dibasic dihydrate, 98+%, for analysis
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate dihydrate,disodium phosphate dihydrate,sodium phosphate dibasic dihydrate,di-sodium hydrogen phosphate dihydrate,phosphoric acid, disodium salt, dihydrate,sodium hydrogenphosphate dihydrate,sodium phosphate, dibasic, dihydrate,disodium hydrogenphosphate dihydrate,dinatrii phosphas dihydricus,2na.hpo4.2h2o PubChem CID: 6451167 IUPAC Name: disodium;hydrogen phosphate;dihydrate SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
Sodium phosphate dibasic dihydrate, specified accord. to the requir. of USP, Ph.Eur.
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L IUPAC Name: disodium dihydrate hydrogen phosphate SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
di-Sodium Hydrogen Phosphate Dihydrate Granulated, Supelco
Physical Form: Granules
Sodium Phosphate, Dibasic, Dihydrate, FCC, 98%, Spectrum™ Chemical
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L IUPAC Name: disodium dihydrate hydrogen phosphate SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
di-Sodium hydrogen phosphate dihydrate, MilliporeSigma™
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate dihydrate,disodium phosphate dihydrate,sodium phosphate dibasic dihydrate,di-sodium hydrogen phosphate dihydrate,phosphoric acid, disodium salt, dihydrate,sodium hydrogenphosphate dihydrate,sodium phosphate, dibasic, dihydrate,disodium hydrogenphosphate dihydrate,dinatrii phosphas dihydricus,2na.hpo4.2h2o PubChem CID: 6451167 IUPAC Name: disodium dihydrate hydrogen phosphate SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
di-Sodium hydrogen phosphate dihydrate, EMSURE™, MilliporeSigma™
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate dihydrate,disodium phosphate dihydrate,sodium phosphate dibasic dihydrate,di-sodium hydrogen phosphate dihydrate,phosphoric acid, disodium salt, dihydrate,sodium hydrogenphosphate dihydrate,sodium phosphate, dibasic, dihydrate,disodium hydrogenphosphate dihydrate,dinatrii phosphas dihydricus,2na.hpo4.2h2o PubChem CID: 6451167 IUPAC Name: disodium dihydrate hydrogen phosphate SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
Sodium Phosphate, Dibasic, Dihydrate, USP, EP, bioCERTIFIED™, 2.5 kg, Spectrum Chemical
CAS: 10028-24-7 Molecular Weight (g/mol): 177.99
Sodium phosphate dibasic dihydrate, meets analytical spec. of BP, Ph. Eur., 98.5-101% (calc. to the dried substance), Solstice
CAS: 10028-24-7 Molecular Formula: H5Na2O6P Molecular Weight (g/mol): 177.99 MDL Number: MFCD00149182 InChI Key: KDQPSPMLNJTZAL-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate dihydrate,disodium phosphate dihydrate,sodium phosphate dibasic dihydrate,di-sodium hydrogen phosphate dihydrate,phosphoric acid, disodium salt, dihydrate,sodium hydrogenphosphate dihydrate,sodium phosphate, dibasic, dihydrate,disodium hydrogenphosphate dihydrate,dinatrii phosphas dihydricus,2na.hpo4.2h2o PubChem CID: 6451167 SMILES: O.O.[Na+].[Na+].OP([O-])([O-])=O
- 1
- 2