accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1003-04-9

IUPAC Name: thiolan-3-one
Synonyms: thiolan-3-one
Molecular Weight (g/mol): 102.15
Molecular Formula: C4H6OS
InChi Key: DSXFPRKPFJRPIB-UHFFFAOYSA-N
SMILES: O=C1CCSC1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)