accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1003-66-3

OD

CAS RN 1003-66-3

IUPAC Name: [(²H)oxy]benzene
Synonyms: [(²H)oxy]benzene
Molecular Weight (g/mol): 95.12
Molecular Formula: C6H5DO
InChi Key: ISWSIDIOOBJBQZ-DYCDLGHISA-N
SMILES: [2H]OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results