Learn More
Phenol-d1, 95% (Isotopic), Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Sensitivity | Air and Light |
| Quantity | 1 g |
| Density | 1.082 |
| Melting Point | 40°C to 42°C |
| Boiling Point | 182°C |
| UN Number | UN1671 |
| Formula Weight | 95.12 |
| Percent Purity | 95% |
| Chemical Name or Material | Phenol-d1, Isotopic |
Safety and Handling
| Hazard Category | H301+H311-H314-H330-H335-H341-H373 |
| Hazard Statement | GHS H Statement H300-H310-H330-H314-H318-H341-H373 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Suspected of causing genetic defects. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENOL, SOLID |
| TSCA | No |
| Recommended Storage | Ambient |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.