accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1003-78-7

IUPAC Name: 2,4-dimethyl-1λ⁶-thiolane-1,1-dione
Synonyms: 2,4-dimethylsulfolane
Molecular Weight (g/mol): 148.22
Molecular Formula: C6H12O2S
InChi Key: WKFQMDFSDQFAIC-UHFFFAOYNA-N
SMILES: CC1CC(C)S(=O)(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results