Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylsulfolane 95%, Thermo Scientific™

Catalog No. AC116830100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC116830100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC116830100 Supplier Thermo Scientific Chemicals Supplier No. 116830100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-78-7
Molecular Formula C6H12O2S
Molecular Weight (g/mol) 148.22
InChI Key WKFQMDFSDQFAIC-UHFFFAOYSA-N
Synonym 2,4-dimethylsulfolane, 2,4-dimethyl sulfolane, 2,4-dimethyltetramethylene sulfone, thiophene, tetrahydro-2,4-dimethyl-, 1,1-dioxide, 2,4-dimethyltetrahydrothiophene dioxide, 2,4-dimethyltetrahydrothiophene 1,1-dioxide, thiophene,tetrahydro-2,4-dimethyl-, 1,1-dioxide, 2,4-dimethyltetrahydrothiophene-1,1-dioxide, 2,4-dimethylthiolane-1,1-dione, dimethylsulfolane
PubChem CID 70483
IUPAC Name 2,4-dimethylthiolane 1,1-dioxide
SMILES CC1CC(S(=O)(=O)C1)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.13
Infrared Spectrum Authentic
Refractive Index 1.4720 to 1.4740 (20°C, 589nm)
Quantity 10 g
Merck Index 15, 3282
Formula Weight 148.22
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethylsulfolane, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.