accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3NHSOOS

CAS RN 100342-30-1

IUPAC Name: N-tert-butylthiophene-2-sulfonamide
Synonyms: N-tert-butylthiophene-2-sulfonamide
Molecular Weight (g/mol): 219.32
Molecular Formula: C8H13NO2S2
InChi Key: CLKMBGGZGFULOO-UHFFFAOYSA-N
SMILES: CC(C)(C)NS(=O)(=O)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results