CAS RN 10035-04-8
Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™
CAS: 10035-04-8 Molecular Formula: Ca Cl2 · 2 H2O
Calcium chloride dihydrate, 99+%, ACS reagent
CAS: 10035-04-8 Molecular Formula: CaCl2·2H2O Molecular Weight (g/mol): 147.02 MDL Number: MFCD00149613
Calcium chloride dihydrate, ACS, 99.0-105.0%
CAS: 10035-04-8 Molecular Formula: CaCl2·2H2O MDL Number: MFCD00149613
Calcium chloride dihydrate, 99% min
CAS: 10035-04-8 Molecular Formula: CaCl2·2H2O MDL Number: MFCD00149613
Calcium Chloride Dihydrate (USP/FCC/EP), Fisher Chemical
CAS: 10035-04-8 Molecular Formula: Ca Cl2 · 2 H2O MDL Number: MFCD00149613
Calcium Chloride Dihydrate (Certified ACS), Fisher Chemical™
CAS: 10035-04-8 Molecular Formula: Ca Cl2 · 2 H2O MDL Number: MFCD00149613
Calcium Chloride, 0.0100 M (M/100), Ricca Chemical
CAS: 10035-04-8
Calcium Chloride Dihydrate, MP Biomedicals™
CAS: 10035-04-8 Molecular Formula: CaCl2. 2H2O
Calcium chloride dihydrate, 99+%, for analysis
CAS: 10035-04-8 Molecular Formula: CaCl2·2H2O Molecular Weight (g/mol): 147.02
Calcium Chloride, Dihydrate, Reagent, ACS, 99-105%, Spectrum™ Chemical
CAS: 10035-04-8 Molecular Formula: CaCl2H4O2 Molecular Weight (g/mol): 147.01 InChI Key: LLSDKQJKOVVTOJ-UHFFFAOYSA-L IUPAC Name: calcium dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ca++]
MilliporeSigma™ Calcium Chloride, Dihydrate, Calbiochem™,
CAS: 10035-04-8 Molecular Formula: CaCl2·2(H2O) Molecular Weight (g/mol): 147.01 Synonym: CaCl2
Calcium chloride, 2.75% w/v aq. soln.
CAS: 10035-04-8 MDL Number: MFCD00010903
CalPlus™ (Brand of Calcium Chloride Dihydrate), Multi-Compendial, U.S.P., J.T. Baker™
CAS: 10035-04-8 Molecular Formula: CaCl2·2H2O