Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride Dihydrate, MP Biomedicals™

Catalog No. MP21995245 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21995245 500 g
MP219952401 1 kg
MP219952405 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP21995245 Supplier MP Biomedicals, Inc Supplier No. 02199524.5
Add to Cart
Add to Cart

Useful in preparing competent bacteria. Calcium Chloride Dihydrate, MP Biomedicals is a USP-grade biochemical reagent. It is used in the preparation of calcium-alginate beads and to compose the medium for Thermophilic Clostridia (MTC). It may be used for

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2. 2H2O

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water with heat liberation, ethanol, acetic acid, acetone.
Formula Weight 147.01
Percent Purity 99.0 to 107.0%
Grade USP
Recommended Storage Store at Room Temperature (15–30°C). Store Desiccated.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.