accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3H3C

CAS RN 1004-66-6

IUPAC Name: 2-methoxy-1,3-dimethylbenzene
Synonyms: 2,6-dimethylanisole
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: GFNZJAUVJCGWLW-UHFFFAOYSA-N
SMILES: COC1=C(C)C=CC=C1C
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)