accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10040-95-6

IUPAC Name: 1-(4-methoxyphenyl)-1H-imidazole
Synonyms: 1-(4-methoxyphenyl)imidazole
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: XNLOIFUGGCCEQX-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1C=CN=C1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results