accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10040-98-9

IUPAC Name: 4-(1H-imidazol-1-yl)benzaldehyde
Synonyms: 4-(imidazol-1-yl)benzaldehyde
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: DCICUQFMCRPKHZ-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)N1C=CN=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)