Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Imidazolyl)benzaldehyde 97.0+%, TCI America™
SDP

Catalog No. I08185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08185G 5 g
I08181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08185G Supplier TCI America Supplier No. I08185G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10040-98-9
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD00209977
InChI Key DCICUQFMCRPKHZ-UHFFFAOYSA-N
Synonym 4-1h-imidazol-1-yl benzaldehyde, 4-imidazol-1-yl-benzaldehyde, 4-1-imidazolyl benzaldehyde, 1-4-formylphenyl imidazole, 4-imidazol-1-yl benzaldehyde, 1-4-formylphenyl-1h-imidazole, 4-1h-imidazol-1-yl-benzaldehyde, benzaldehyde, 4-1h-imidazol-1-yl, 4-1h-imidazol-1-yl benzenecarbaldehyde, p-1-imidazolyl benzaldehyde
PubChem CID 736530
IUPAC Name 4-imidazol-1-ylbenzaldehyde
SMILES C1=CC(=CC=C1C=O)N2C=CN=C2

Specifications

Melting Point 149°C
Quantity 5 g
Formula Weight 172.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1-Imidazolyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.