accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10041-06-2

H3CONN

CAS RN 10041-06-2

IUPAC Name: 1-[4-(1H-imidazol-1-yl)phenyl]ethan-1-one
Synonyms: 1-[4-(imidazol-1-yl)phenyl]ethanone
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: GAIQQJIMVVUTQN-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)N1C=CN=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results