Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-(1-Imidazolyl)acetophenone, 97%, Thermo Scientific™

Catalog No. AAL0611506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0611506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0611506 Supplier Thermo Scientific Chemicals Supplier No. L0611506

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10041-06-2
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD00005282
InChI Key GAIQQJIMVVUTQN-UHFFFAOYSA-N
Synonym 4-imidazol-1-yl acetophenone, 4'-imidazol-1-yl acetophenone, 1-4-1h-imidazol-1-yl phenyl ethanone, 4'-1-imidazolyl acetophenone, 1-4-imidazol-1-yl-phenyl-ethanone, 1-4-acetylphenyl imidazole, ethanone, 1-4-1h-imidazol-1-yl phenyl, 1-4-imidazol-1-ylphenyl ethanone, 1-4-1h-imidazol-1-yl phenyl ethan-1-one, 4'-1h-imidazol-1-yl acetophenone
PubChem CID 82316
IUPAC Name 1-(4-imidazol-1-ylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)N2C=CN=C2

Specifications

Melting Point 114°C to 118°C
Quantity 5 g
Beilstein 608955
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4'-(1-Imidazolyl)acetophenone
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 233-123-5
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.