accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100478-25-9

IUPAC Name: 4,6-bis(difluoromethoxy)-2-(methylsulfanyl)pyrimidine
Synonyms: 4,6-bis(difluoromethoxy)-2-(methylsulfanyl)pyrimidine
Molecular Weight (g/mol): 258.19
Molecular Formula: C7H6F4N2O2S
InChi Key: HURDQHYOXAOOJJ-UHFFFAOYSA-N
SMILES: CSC1=NC(OC(F)F)=CC(OC(F)F)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results