CAS RN 1005771-04-9
CAS RN 1005771-04-9
IUPAC Name:
1,3-dibromo-7-tert-butylpyrene
Synonyms:
1,3-dibromo-7-tert-butylpyrene
Molecular Weight (g/mol):
416.16
Molecular Formula:
C20H16Br2
InChi Key:
BARYQDOQFLJABV-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC2=CC=C3C(Br)=CC(Br)=C4C=CC(=C1)C2=C34
1
–
1
of
1
results
1
1,3-Dibromo-7-tert-butylpyrene 97.0+%, TCI America™

CAS: 1005771-04-9 Molecular Formula: C20H16Br2 Molecular Weight (g/mol): 416.156 InChI Key: BARYQDOQFLJABV-UHFFFAOYSA-N PubChem CID: 57953468 IUPAC Name: 1,3-dibromo-7-tert-butylpyrene SMILES: CC(C)(C)C1=CC2=C3C(=C1)C=CC4=C3C(=C(C=C4Br)Br)C=C2