Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromo-7-tert-butylpyrene 97.0+%, TCI America™
SDP

Catalog No. D41875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41875G 5 g
D41871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41875G Supplier TCI America Supplier No. D41875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1005771-04-9
Molecular Formula C20H16Br2
Molecular Weight (g/mol) 416.156
InChI Key BARYQDOQFLJABV-UHFFFAOYSA-N
PubChem CID 57953468
IUPAC Name 1,3-dibromo-7-tert-butylpyrene
SMILES CC(C)(C)C1=CC2=C3C(=C1)C=CC4=C3C(=C(C=C4Br)Br)C=C2

Specifications

Quantity 5 g
Formula Weight 416.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromo-7-tert-butylpyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.