accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOOOOOOOGdGd

CAS RN 100655-00-3

IUPAC Name: tris(oxalic acid) digadolinium hydrate
Synonyms: tris(oxalic acid) digadolinium hydrate
Molecular Weight (g/mol): 602.62
Molecular Formula: C6H8Gd2O13
InChi Key: FEIDMMDEDUAFJQ-UHFFFAOYSA-N
SMILES: O.[Gd].[Gd].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results