accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1009-61-6

IUPAC Name: 1-(4-acetylphenyl)ethan-1-one
Synonyms: P-acetylacetophenone
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: SKBBQSLSGRSQAJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)